7-Fluoronaphthalen-2-ol ≥95%

Supplier: Ambeed
Warning

A807954-250MG A807954-100MG
77622-966EA 141.11 USD
77622-966 77784-105
7-Fluoronaphthalen-2-ol ≥95%
7-Fluoronaphthalen-2-ol

Precaución: for Research Use Only

Formula: C₁₀H₇FO
MDL Number: MFCD17012580
CAS Number: 889884-94-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR