7-Oxa-4-azaspiro[2.5]octan-5-one ≥95%

Supplier: Ambeed
Warning

A807666-100MG A807666-1G A807666-250MG
77886-792EA 199 USD
77886-792 77886-793 77886-794
7-Oxa-4-azaspiro[2.5]octan-5-one ≥95%
7-Oxa-4-azaspiro[2.5]octan-5-one

Precaución: for Research Use Only

Formula: C₆H₉NO₂
MDL Number: MFCD11655976
CAS Number: 1100753-07-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR