[1,1'-Bi(cyclohexane)]-1-carboxylic acid ≥98%, Ambeed.Inc

Supplier: Ambeed
Warning

A807269-100MG A807269-1G A807269-250MG A807269-5G
77684-756EA 146 USD
77684-756 77684-757 77684-758 77893-588
[1,1'-Bi(cyclohexane)]-1-carboxylic acid ≥98%, Ambeed.Inc
[1,1'-Bi(cyclohexane)]-1-carboxylic acid

Precaución: For research use only.

Formula: C₁₃H₂₂O₂
MDL Number: MFCD00129987
CAS Number: 60263-54-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR