tert-Butyl 7-(hydroxymethyl)-2-azaspiro[3.5]nonane-2-carboxylate ≥95%

Supplier: Ambeed
Warning

A806869-100MG A806869-1G A806869-5G
77686-832EA 74 USD
77686-832 77686-833 77901-607
tert-Butyl 7-(hydroxymethyl)-2-azaspiro[3.5]nonane-2-carboxylate ≥95%
tert-Butyl 7-(hydroxymethyl)-2-azaspiro[3.5]nonane-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₂₅NO₃
MDL Number: MFCD27956901
CAS Number: 1434141-69-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR