5-(Hydroxymethyl)-1H-benzimidazole ≥98%

Supplier: Ambeed
Warning

A805788-1G A805788-100MG A805788-250MG
77573-580EA 43 USD
77573-580 77670-142 77670-144
5-(Hydroxymethyl)-1H-benzimidazole ≥98%
5-(Hydroxymethyl)-1H-benzimidazole

Precaución: for Research Use Only

Formula: C₈H₈N₂O
MDL Number: MFCD08690267
CAS Number: 106429-29-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR