(R,Z)-2-(Oleoyloxy)-3-(palmitoyloxy)propyl (2-(trimethylammonio)ethyl) phosphate ≥98%

Supplier: Ambeed
Warning

Synonyms: 1-Palmitoyl-2-oleoyl-sn-glycero-3-PC

A805109-100MG A805109-250MG
77747-551EA 127 USD
77747-551 77747-552
(R,Z)-2-(Oleoyloxy)-3-(palmitoyloxy)propyl (2-(trimethylammonio)ethyl) phosphate ≥98%
(R,Z)-2-(Oleoyloxy)-3-(palmitoyloxy)propyl (2-(trimethylammonio)ethyl) phosphate

Precaución: for Research Use Only

Formula: C₄₂H₈₂NO₈P
MW: 760.08 g/mol
MDL Number: MFCD00043212
CAS Number: 26853-31-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR