(4-Chlorophenyl)acetyl chloride ≥98%

Supplier: Ambeed
Danger

Synonyms: 2-(4-Chlorophenyl)acetyl chloride, 4-Chlorobenzeneacetyl chloride

A805058-25G A805058-5G
77573-578EA 27.81 USD
77573-578 77674-180
(4-Chlorophenyl)acetyl chloride ≥98%
(4-Chlorophenyl)acetyl chloride

Precaución: for Research Use Only

Formula: (C₆H₄Cl)CH₂COCl
MW: 189.04 g/mol
MDL Number: MFCD00037111
CAS Number: 25026-34-0
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR