4-Chloro-3-nitro-tert-butylbenzene ≥95%

Supplier: Ambeed
Warning

A804019-25G A804019-5G A804019-10G A804019-1G
77878-647EA 228 USD
77878-647 77878-648 77878-645 77878-646
4-Chloro-3-nitro-tert-butylbenzene ≥95%
4-Chloro-3-nitro-tert-butylbenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂ClNO₂
MDL Number: MFCD07368382
CAS Number: 58574-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR