(R)-2-(1,3-dioxoisoindolin-2-yl)propanoic acid ≥95%

Supplier: Ambeed
Warning

A803358-1G A803358-250MG
77622-962EA 136.99 USD
77622-962 77622-964
(R)-2-(1,3-dioxoisoindolin-2-yl)propanoic acid ≥95%
(R)-2-(1,3-dioxoisoindolin-2-yl)propanoic acid

Precaución: for Research Use Only

Formula: C₁₁H₉NO₄
MDL Number: MFCD24479562
CAS Number: 29588-83-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR