1,4-Dibromophthalazine ≥95%

Supplier: Ambeed
Warning

A802914-1G A802914-5G
77599-090EA 89.18 USD
77599-090 77784-096
1,4-Dibromophthalazine ≥95%
1,4-Dibromophthalazine

Precaución: for Research Use Only

Formula: C₈H₄Br₂N₂
MW: 287.9388 g/mol
MDL Number: MFCD13659422
CAS Number: 3660-90-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR