Bicyclo[2.2.1]hept-5-en-2-one ≥95%

Supplier: Ambeed
Warning

A802356-1G A802356-5G
77573-574EA 32.96 USD
77573-574 77595-278
Bicyclo[2.2.1]hept-5-en-2-one ≥95%
Bicyclo[2.2.1]hept-5-en-2-one

Precaución: for Research Use Only

Formula: C₇H₈O
MW: 108.13 g/mol
MDL Number: MFCD00101751
CAS Number: 694-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR