4-Bromo-6-fluoro-1,3-benzoxazole ≥97%

Supplier: Ambeed
Warning

A802191-100MG A802191-250MG
77749-587EA 42 USD
77749-587 77749-588
4-Bromo-6-fluoro-1,3-benzoxazole ≥97%
4-Bromo-6-fluoro-1,3-benzoxazole

Precaución: for Research Use Only

Formula: C₇H₃BrFNO
MDL Number: MFCD29049291
CAS Number: 1929606-78-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR