p-Terphenyl ≥98%

Supplier: Ambeed
Danger

Synonyms: 4-Phenyl-1,1'-biphenyl, 1,4-Diphenylbenzene, 4-Phenylbiphenyl, 1,4-Terphenyl

A801812-25G A801812-500G
77573-566EA 25.75 USD
77573-566 77749-584
p-Terphenyl ≥98%
p-Terphenyl

Precaución: for Research Use Only

Formula: C₁₈H₁₄
MW: 230.31 g/mol
MDL Number: MFCD00003061
CAS Number: 92-94-4
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR