(2-Chloro-3-methoxypyridin-4-yl)boronic acid ≥95%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-3-methoxypyridine-4-boronic acid

A801091-250MG A801091-100MG A801091-1G A801091-10G A801091-5G
77784-073EA 118 USD
77784-073 77784-070 77784-072 77784-071 77784-074
(2-Chloro-3-methoxypyridin-4-yl)boronic acid ≥95%
(2-Chloro-3-methoxypyridin-4-yl)boronic acid

Precaución: for Research Use Only

Formula: C₆H₇BClNO₃
MDL Number: MFCD16610616
CAS Number: 2096339-60-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR