6-Phenyl-2,2'-bipyridine ≥98%

Supplier: Ambeed
Warning

A801005-250MG A801005-1G A801005-5G
77595-274EA 164 USD
77595-274 77622-950 77784-069
6-Phenyl-2,2'-bipyridine ≥98%
6-Phenyl-2,2'-bipyridine

Precaución: for Research Use Only

Formula: C₁₆H₁₂N₂
MW: 232.28 g/mol
MDL Number: MFCD28053905
CAS Number: 61633-06-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR