Benzo[d][1,2,3]thiadiazol-5-amine ≥98%

Supplier: Ambeed
Warning

A800997-1G A800997-100MG A800997-250MG A800997-50MG
77674-166EA 810 USD
77674-166 77884-741 77884-742 77884-743
Benzo[d][1,2,3]thiadiazol-5-amine ≥98%
Benzo[d][1,2,3]thiadiazol-5-amine

Precaución: for Research Use Only

Formula: C₆H₅N₃S
MDL Number: MFCD20690748
CAS Number: 1753-29-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR