Methyl cis-11-eicosenoate ≥98%

Supplier: Ambeed
Warning

A800700-1G A800700-250MG A800700-5G
77627-588EA 538.69 USD
77627-588 77627-590 77878-641
Methyl cis-11-eicosenoate ≥98%
Methyl cis-11-eicosenoate

Precaución: for Research Use Only

Formula: C₂₁H₄₀O₂
MDL Number: MFCD00010458
CAS Number: 2390-09-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR