1,3,5-Tri(1H-pyrazol-4-yl)benzene ≥95%

Supplier: Ambeed
Warning

A800090-1G A800090-250MG
77573-556EA 274 USD
77573-556 77784-062
1,3,5-Tri(1H-pyrazol-4-yl)benzene ≥95%
1,3,5-Tri(1H-pyrazol-4-yl)benzene

Precaución: for Research Use Only

Formula: C₁₅H₁₂N₆
MW: 276.296 g/mol
MDL Number: MFCD30741245
CAS Number: 1325728-09-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR