Methyl 7-oxoheptanoate ≥95%

Supplier: Ambeed
Warning

A799902-1G A799902-250MG A799902-5G
77599-078EA 370.03 USD
77599-078 77599-080 77622-946
Methyl 7-oxoheptanoate ≥95%
Methyl 7-oxoheptanoate

Precaución: for Research Use Only

Formula: C₈H₁₄O₃
MW: 158.2 g/mol
MDL Number: MFCD00055616
CAS Number: 35376-00-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR