2-Aminobutanenitrile hydrochloride ≥95%

Supplier: Ambeed
Warning

A799897-100MG A799897-250MG
77784-056EA 85 USD
77784-056 77784-057
2-Aminobutanenitrile hydrochloride ≥95%
2-Aminobutanenitrile hydrochloride

Precaución: for Research Use Only

Formula: C₄H₉ClN₂
MDL Number: MFCD11858092
CAS Number: 93554-80-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR