2-((2,6-Dimethylphenyl)amino)-N,N-diethyl-2-oxoethan-1-amine oxide ≥98%

Supplier: Ambeed
Warning

A799762-100MG A799762-250MG
77622-944EA 76.22 USD
77622-944 77784-054
2-((2,6-Dimethylphenyl)amino)-N,N-diethyl-2-oxoethan-1-amine oxide ≥98%
2-((2,6-Dimethylphenyl)amino)-N,N-diethyl-2-oxoethan-1-amine oxide

Precaución: for Research Use Only

Formula: C₁₄H₂₂N₂O₂
MDL Number: MFCD22125322
CAS Number: 2903-45-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR