Ethyl 4,4-difluoro-2-(ethoxymethylene)acetoacetate ≥98%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-(ethoxymethylene)-4,4-difluoro-3-oxobutanoate, Ethyl 4,4-difluoro-2-(ethoxymethylene)-3-oxobutanoate, Ethyl 4,4-difluoro-2-(ethoxymethylidene)-3-oxobutanoate

A799108-1G A799108-5G
77684-742EA 152 USD
77684-742 77886-784
Ethyl 4,4-difluoro-2-(ethoxymethylene)acetoacetate ≥98%, Ambeed.Inc
Ethyl 4,4-difluoro-2-(ethoxymethylene)acetoacetate

Precaución: For research use only.

Formula: C₉H₁₂F₂O₄
MDL Number: MFCD10567161
CAS Number: 176969-33-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR