(6-(Methylcarbamoyl)pyridin-3-yl)boronic acid ≥98%

Supplier: Ambeed
Warning

A799038-100MG A799038-1G A799038-250MG A799038-5G
77886-780EA 114 USD
77886-780 77886-781 77886-782 77886-783
(6-(Methylcarbamoyl)pyridin-3-yl)boronic acid ≥98%
(6-(Methylcarbamoyl)pyridin-3-yl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₉BN₂O₃
MDL Number: MFCD08458474
CAS Number: 1217340-94-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR