3'-Fluorobiphenyl-4-carboxylic acid ≥95%

Supplier: Ambeed
Warning

A797244-100MG A797244-250MG A797244-5G
77893-570EA 30 USD
77893-570 77893-571 77893-572
3'-Fluorobiphenyl-4-carboxylic acid ≥95%
3'-Fluorobiphenyl-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₃H₉FO₂
MDL Number: MFCD07779362
CAS Number: 137045-30-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR