1-Dodecanol ≥98%

Supplier: Ambeed
Danger

Synonyms: Undecyl carbinol, 1-Dodecyl alcohol, 1-Hydroxy dodecane, Lauryl alcohol, Dodecyl Alcohol

A796596-100G A796596-500G
77573-550EA 5 USD
77573-550 77618-216
1-Dodecanol ≥98%
1-Dodecanol

Precaución: for Research Use Only

Formula: H₃C(CH₂)₁₀CH₂OH
MW: 186.34 g/mol
Boiling Pt: 260
Flash Pt: 119
MDL Number: MFCD00004753
CAS Number: 112-53-8
UN: 3077
ADR: 9 Ⅲ
Merck Index: 13,03439

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR