4-(N,N-Dimethylsulfamoyl)benzeneboronic acid, pinacol ester ≥98%

Supplier: Ambeed
Warning

Synonyms: 4-[(Dimethylamino)sulphonyl]benzeneboronic acid, pinacol ester, N,N-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzenesulphonamide

A796524-1G A796524-250MG A796524-25G
77614-422EA 83.43 USD
77614-422 77882-715 77882-716
4-(N,N-Dimethylsulfamoyl)benzeneboronic acid, pinacol ester ≥98%
4-(N,N-Dimethylsulfamoyl)benzeneboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₂₂BNO₄S
MDL Number: MFCD03425962
CAS Number: 486422-04-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR