(1S,2R)-2-methylcyclopropane-1-carboxylic acid ≥97%

Supplier: Ambeed
Danger

A795457-100MG A795457-1G A795457-250MG
77749-567EA 250 USD
77749-567 77749-568 77749-569
(1S,2R)-2-methylcyclopropane-1-carboxylic acid ≥97%
(1S,2R)-2-methylcyclopropane-1-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₈O₂
MW: 100.12 g/mol
MDL Number: MFCD19238068
CAS Number: 26510-98-5
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR