(2-(N-Methylsulfamoyl)phenyl)boronic acid ≥98%

Supplier: Ambeed
Warning

A794360-1G A794360-250MG A794360-5G
77891-539EA 53 USD
77891-539 77891-540 77891-541
(2-(N-Methylsulfamoyl)phenyl)boronic acid ≥98%
(2-(N-Methylsulfamoyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₁₀BNO₄S
MDL Number: MFCD09038342
CAS Number: 956283-09-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR