[2,2'-Bithiophene]-5,5'-diyldiboronic acid ≥95%

Supplier: Ambeed
Warning

A794231-100MG A794231-1G A794231-250MG A794231-5G
77886-767EA 27 USD
77886-767 77886-768 77886-769 77886-770
[2,2'-Bithiophene]-5,5'-diyldiboronic acid ≥95%
[2,2'-Bithiophene]-5,5'-diyldiboronic acid

Precaución: for Research Use Only

Formula: C₈H₈B₂O₄S₂
MDL Number: MFCD11504864
CAS Number: 189358-30-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR