rel-(1R,2R)-2-(4-Phenylpiperidin-1-yl)cyclohexan-1-ol hydrochloride ≥99%

Supplier: Ambeed
Warning

A793614-10MG A793614-1MG A793614-5MG
77618-208EA 72.43 USD
77618-208 77618-210 77618-212
rel-(1R,2R)-2-(4-Phenylpiperidin-1-yl)cyclohexan-1-ol hydrochloride ≥99%
rel-(1R,2R)-2-(4-Phenylpiperidin-1-yl)cyclohexan-1-ol hydrochloride

Precaución: for Research Use Only

Formula: C₁₇H₂₆ClNO
MDL Number: MFCD06804635
CAS Number: 120447-62-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR