3-Cyano-1H-indole-5-carboxylic acid ≥95%

Supplier: Ambeed
Warning

A793179-250MG A793179-100MG A793179-1G
77674-156EA 33 USD
77674-156 77873-256 77873-257
3-Cyano-1H-indole-5-carboxylic acid ≥95%
3-Cyano-1H-indole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₆N₂O₂
MDL Number: MFCD07776627
CAS Number: 889942-87-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR