1-(Benzyloxy)-5-bromo-2,4-difluorobenzene ≥95%
Supplier: Ambeed
|
Warning
|
A793084-1G
A793084-5G
A793084-250MG
77784-031EA
207
USD
77784-031
77784-033
77784-032
1-(Benzyloxy)-5-bromo-2,4-difluorobenzene ≥95%
1-(Benzyloxy)-5-bromo-2,4-difluorobenzene
Precaución: for Research Use Only
Formula:
C₁₃H₉BrF₂O |
MDL Number:
MFCD29043945 CAS Number: 1881293-65-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...