Bis(pentamethylcyclopentadienyl)ruthenium(II) ≥98%

Supplier: Ambeed
Warning

Synonyms: Bis(pentamethylcyclopentadienyl)ruthenium, Bis(pentamethylcyclopentadienyl)ruthenium(II), Decamethylruthenocene

A792946-100MG A792946-1G A792946-250MG
77614-416EA 39.14 USD
77614-416 77614-418 77684-736
Bis(pentamethylcyclopentadienyl)ruthenium(II) ≥98%
Bis(pentamethylcyclopentadienyl)ruthenium (II)

Precaución: For research use only.

Formula: C₂₀H₃₀Ru
MW: 371.522 g/mol
MDL Number: MFCD00058889
CAS Number: 84821-53-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR