5,5',7-Trihydroxy-2',2'-dimethyl-3-(3-methylbut-2-en-1-yl)-2'H,4H-[2,8'-bichromen]-4-one ≥97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Kuwanon A
A792718-5MG
A792718-2MG
77749-563EA
303
USD
77749-563
77749-562
5,5',7-Trihydroxy-2',2'-dimethyl-3-(3-methylbut-2-en-1-yl)-2'H,4H-[2,8'-bichromen]-4-one ≥97%
5,5',7-Trihydroxy-2',2'-dimethyl-3-(3-methylbut-2-en-1-yl)-2'H,4H-[2,8'-bichromen]-4-one
Precaución: for Research Use Only
Formula:
C₂₅H₂₄O₆ MW: 420.45 g/mol |
MDL Number:
MFCD31382182 CAS Number: 62949-77-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...