4-(4-Fluorophenoxy)-3-methylaniline ≥98%

Supplier: Ambeed
Danger

A792230-1G A792230-5G
77882-704EA 53 USD
77882-704 77882-705
4-(4-Fluorophenoxy)-3-methylaniline ≥98%
4-(4-Fluorophenoxy)-3-methylaniline

Precaución: for Research Use Only

Formula: C₁₃H₁₂FNO
MDL Number: MFCD08687897
CAS Number: 946663-67-8
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR