N-(2-Amino-5-(thiophen-2-yl)phenyl)-2-(piperazin-1-yl)quinoline-6-carboxamide ≥99%
Supplier: Ambeed
|
Warning
|
Synonyms:
ACY-957
A792202-1MG
A792202-10MG
A792202-5MG
77618-206EA
27.81
USD
77618-206
77747-542
77747-543
N-(2-Amino-5-(thiophen-2-yl)phenyl)-2-(piperazin-1-yl)quinoline-6-carboxamide ≥99%
N-(2-Amino-5-(thiophen-2-yl)phenyl)-2-(piperazin-1-yl)quinoline-6-carboxamide
Precaución: for Research Use Only
Formula:
C₂₄H₂₃N₅OS MW: 429.54 g/mol |
MDL Number:
MFCD30342405 CAS Number: 1609389-52-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...