(5-Propionylthiophen-2-yl)boronic acid ≥95%

Supplier: Ambeed
Warning

A791245-100MG A791245-1G A791245-250MG
77891-534EA 37 USD
77891-534 77891-535 77891-536
(5-Propionylthiophen-2-yl)boronic acid ≥95%
(5-Propionylthiophen-2-yl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₉BO₃S
MDL Number: MFCD09952062
CAS Number: 352525-99-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR