neo-Decanoic acid ≥97% (mixture of isomers)

Supplier: Ambeed
Warning

A790990-100G A790990-10G A790990-1G A790990-25G A790990-5G
77880-576EA 30 USD
77880-576 77880-577 77880-578 77880-579 77880-580
neo-Decanoic acid ≥97% (mixture of isomers)
neo-Decanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₂₀O₂
MDL Number: MFCD00068291
CAS Number: 26896-20-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR