N-(4-((1,4,8,11-Tetraazacyclotetradecan-1-yl)methyl)benzyl)-1-(pyridin-2-yl)methanamine hexahydrobromide ≥98%

Supplier: Ambeed
Warning

Synonyms: N-[[4-(1,4,8,11-Tetraazacyclotetradec-1-ylmethyl)phenyl]methyl]-2-pyridinemethanaminehexahydrobromide, AMD 3465 6HBr, AMD 3465 hexahydrobromide, GENZ-644494 hexahydrobromide

A790755-100MG A790755-10MG A790755-5MG
77686-812EA 313 USD
77686-812 77686-813 77686-814
N-(4-((1,4,8,11-Tetraazacyclotetradecan-1-yl)methyl)benzyl)-1-(pyridin-2-yl)methanamine hexahydrobromide ≥98%
N-(4-((1,4,8,11-Tetraazacyclotetradecan-1-yl)methyl)benzyl)-1-(pyridin-2-yl)methanamine hexahydrobromide

Precaución: for Research Use Only

Formula: C₂₄H₄₄Br₆N₆
MW: 896.07 g/mol
MDL Number: MFCD20926342
CAS Number: 185991-07-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR