Dicyclohexyl(2-(2-methoxynaphthalen-1-yl)phenyl)phosphine ≥98%

Supplier: Ambeed
Warning

A790613-250MG A790613-100MG A790613-1G A790613-5G
77618-202EA 248.23 USD
77618-202 77618-198 77618-200 77747-539
Dicyclohexyl(2-(2-methoxynaphthalen-1-yl)phenyl)phosphine ≥98%
Dicyclohexyl(2-(2-methoxynaphthalen-1-yl)phenyl)phosphine

Precaución: for Research Use Only

Formula: C₂₉H₃₅OP
MDL Number: MFCD23704405
CAS Number: 1309570-98-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR