DL-Homocysteine ≥98%

Supplier: Ambeed
Danger

Synonyms: (±)-Homocysteine, 2-Amino-4-mercaptobutanoic acid, H-DL-HoCys-OH

A789113-1G A789113-250MG A789113-5G
77614-412EA 94.76 USD
77614-412 77674-146 77882-699
DL-Homocysteine ≥98%
DL-Homocysteine

Precaución: for Research Use Only

Formula: C₄H₉NO₂S
MW: 135.19 g/mol
MDL Number: MFCD00004898
CAS Number: 454-29-5
UN: 3077
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR