N3-Cyclopropyl-7-(4-isopropylbenzyl)-7H-pyrrolo[3,2-f]quinazoline-1,3-diamine dihydrochloride ≥98%

Supplier: Ambeed
Warning

Synonyms: SCH79797 2HCl

A788997-10MG A788997-5MG
77622-932EA 147.29 USD
77622-932 77622-934
N3-Cyclopropyl-7-(4-isopropylbenzyl)-7H-pyrrolo[3,2-f]quinazoline-1,3-diamine dihydrochloride ≥98%
N3-Cyclopropyl-7-(4-isopropylbenzyl)-7H-pyrrolo[3,2-f]quinazoline-1,3-diamine dihydrochloride

Precaución: for Research Use Only

Formula: C₂₃H₂₇Cl₂N₅
MW: 444.4 g/mol
MDL Number: MFCD04039788
CAS Number: 1216720-69-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR