7-Bromo-2,4-dichloroquinoline ≥95%

Supplier: Ambeed
Warning

A788965-1G A788965-100MG A788965-250MG
77595-254EA 301.35 USD
77595-254 77749-554 77749-555
7-Bromo-2,4-dichloroquinoline ≥95%
7-Bromo-2,4-dichloroquinoline

Precaución: for Research Use Only

Formula: C₉H₄BrCl₂N
MW: 276.94 g/mol
MDL Number: MFCD22200006
CAS Number: 406205-10-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR