4-Methoxybenzo[d]oxazole ≥97%

Supplier: Ambeed
Warning

A788839-1G A788839-250MG
77749-552EA 353 USD
77749-552 77749-553
4-Methoxybenzo[d]oxazole ≥97%
4-Methoxybenzo[d]oxazole

Precaución: for Research Use Only

Formula: C₈H₇NO₂
MW: 149.15 g/mol
MDL Number: MFCD13176880
CAS Number: 192753-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR