3,5-Difluoro-4-methoxybenzeneboronic acid pinacol ester ≥95%

Supplier: Ambeed
Warning

Synonyms: 2-(3,5-Difluoro-4-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3,5-Difluoro-4-methoxyphenylboronic acid pinacol ester

A787011-1G A787011-5G A787011-250MG
77784-024EA 208 USD
77784-024 77784-026 77784-025
3,5-Difluoro-4-methoxybenzeneboronic acid pinacol ester ≥95%
3,5-Difluoro-4-methoxybenzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₇BF₂O₃
MDL Number: MFCD22414481
CAS Number: 890839-37-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR