Fmoc-3,4-dimethoxy-L-phenylalanine ≥98%

Supplier: Ambeed
Warning

Synonyms: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(3,4-dimethoxyphenyl)propanoic acid, Fmoc-Phe(3,4-DiOMe)-OH

A786780-100MG A786780-250MG
77880-567EA 27 USD
77880-567 77880-568
Fmoc-3,4-dimethoxy-L-phenylalanine ≥98%
Fmoc-3,4-dimethoxy-L-phenylalanine

Precaución: for Research Use Only

Formula: C₂₆H₂₅NO₆
MW: 447.49 g/mol
MDL Number: MFCD01317730
CAS Number: 184962-88-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR