(E)-N-Benzylidene-1-phenylmethanamine ≥95%

Supplier: Ambeed
Warning

A786423-5G A786423-1G
77670-106EA 54 USD
77670-106 77880-565
(E)-N-Benzylidene-1-phenylmethanamine ≥95%
(E)-N-Benzylidene-1-phenylmethanamine

Precaución: for Research Use Only

Formula: C₁₄H₁₃N
CAS Number: 27845-50-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR