1-(4-Methylquinazolin-2-yl)guanidine hydrochloride ≥95%

Supplier: Ambeed
Warning

A785893-100MG A785893-10MG A785893-5MG
77686-804EA 125 USD
77686-804 77686-805 77686-806
1-(4-Methylquinazolin-2-yl)guanidine hydrochloride ≥95%
1-(4-Methylquinazolin-2-yl)guanidine hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₂ClN₅
MDL Number: MFCD00035249
CAS Number: 5361-15-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR