(R)-5-Chloro-N-((2-oxo-3-(4-(3-oxomorpholino)phenyl)oxazolidin-5-yl)methyl)thiophene-2-carboxamide ≥99%

Supplier: Ambeed
Warning

Synonyms: 5-R-Rivaroxaban

A785317-100MG A785317-1G A785317-250MG
77618-170EA 80.34 USD
77618-170 77618-172 77618-174
(R)-5-Chloro-N-((2-oxo-3-(4-(3-oxomorpholino)phenyl)oxazolidin-5-yl)methyl)thiophene-2-carboxamide ≥99%
(R)-5-Chloro-N-((2-oxo-3-(4-(3-oxomorpholino)phenyl)oxazolidin-5-yl)methyl)thiophene-2-carboxamide

Precaución: for Research Use Only

Formula: C₁₉H₁₈ClN₃O₅S
MW: 435.88 g/mol
MDL Number: MFCD17171369
CAS Number: 865479-71-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR