Maduramicin ammonium ≥98%

Supplier: Ambeed
Danger

Synonyms: Cygro, Ammonium 2-((2R,3S,4S,5R,6S)-6-((R)-1-((2S,5R,7S,8R,9S)-2-((2S,2'R,3'S,5R,5'R)-3'-(((2R,4S,5S,6S)-4,5-dimethoxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-5'-((2S,3S,5R,6S)-6-hydroxy-3,5,6-trimethyltetrahydro-2H-pyran-2-yl)-2-methyloctahydro-[2,2'-bifuran]-5-yl)-9-hydroxy-2,8-dimethyl-1,6-dioxaspiro[4.5]decan-7-yl)ethyl)-2-hydroxy-4,5-dimethoxy-3-methyltetrahydro-2H-pyran-2-yl)acetate, Azane, 2-[(2R,3S,4S,5R,6S)-6-[(1R)-1-[(2S,5R,7S,8R,9S)-2-[(2R,5S)-5-[(2R,3S,5R)-3-[(2R,4S,5S,6S)-4,5-dimethoxy-6-methyloxan-2-yl]oxy-5-[(2S,3S,5R,6S)-6-hydroxy-3,5,6-trimethyloxan-2-yl]oxolan-2-yl]-5-methyloxolan-2-yl]-7-hydroxy-2,8-dimethyl-1,10-dioxaspiro[4.5]decan-9-yl]ethyl]-2-hydroxy-4,5-dimethoxy-3-methyloxan-2-yl]acetic acid, Maduramicin ammonium, Prinicin ammonium

A785107-1MG A785107-5MG
77603-016EA 5.13 USD
77603-016 77603-018
Maduramicin ammonium ≥98%
Maduramicin ammonium

Precaución: for Research Use Only

Formula: C₄₇H₈₃NO₁₇
MW: 934.16 g/mol
MDL Number: MFCD00865282
CAS Number: 84878-61-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR